4-Chlorobenzonitrile 99%

Supplier: Thermo Fisher Scientific
Warning

151871000 151870250 151875000
ACRO151871000EA 94.2 EUR
ACRO151871000 ACRO151870250 ACRO151875000
4-Chlorobenzonitrile 99%
4-Chlorobenzonitrile
Formula: C₇H₄ClN
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 91…94 °C
Density: 1,1133 g/cm³ (16.8 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4

Order Now

Specification Test Results

Appearance White to yellow Flakes and/or chunks
Infrared spectrum Conforms
Melting point 91 to 94 °C
Water ≤0.1 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR