Trichloroacetic acid (TCA) 99%, extra pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: TCA

152130010 152135000 152130025 152130100
ACRO152130010EA 172 EUR
ACRO152130010 ACRO152135000 ACRO152130025 ACRO152130100
Trichloroacetic acid (TCA) 99%, extra pure
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C (760 mmHg)
Melting Pt: 52…58 °C
Density: 1,62 g/cm³
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8,II
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 52 to 58 °C
Water ≤0.5 % (K.F.)
Titration with NaOH ≥98.5 %
Iron (Fe) ≤10 ppm
Sulfuric acid ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR