2,6-Dichlorophenolindophenol sodium salt hydrate 98+%

Supplier: Thermo Fisher Scientific

152870010 152870100 152870250 152871000
ACRO152870010EA 29.5 EUR
ACRO152870010 ACRO152870100 ACRO152870250 ACRO152871000
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
Melting Pt: ? 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150014
CAS Number: 1266615-56-8
EINECS: 210-640-4

Order Now

Specification Test Results

Appearance Dark green to dark blue-purple Powder
Infrared spectrum Conforms
Loss on drying ≤14 % (1 g, 120°C)
Titration with HClO4 ≥98.0 % (On dry substance)
Suitability (for Ascorbic acid det.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR