Undec-10-enoic acid 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Undecylenic-10 acid, Undecenoic acid, 10-Undecenoic acid, Undecylenic acid, Undecenylic acid

154492500 154495000 154490025
ACRO154492500EA 44.2 EUR
ACRO154492500 ACRO154495000 ACRO154490025
Undec-10-enoic acid 99%
Undec-10-enoic acid
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 275 °C (760 mmHg)
Melting Pt: 24.5 °C
Density: 0.911 g/cm³
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00004442
CAS Number: 112-38-9
EINECS: 203-965-8
Merck Index: 12,09983

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid or liquid
Infrared spectrum Conforms
GC ≥97.0 %
Titration with NaOH 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR