1-Chlorobutane 99+%, pure

Supplier: Thermo Fisher Scientific
Danger

154630010 154630025 154630100
ACRO154630010EA 62.5 EUR
ACRO154630010 ACRO154630025 ACRO154630100
1-Chlorobutane 99+%, pure
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
Boiling Pt: 77…78 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0,88 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3,II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4005 to 1.4035 (20°C, 589 nm)
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR