Citraconic anhydride 98%

Supplier: Thermo Fisher Scientific
Danger

154791000 154792500 154790010 154790250
ACRO154791000EA 61.9 EUR
ACRO154791000 ACRO154792500 ACRO154790010 ACRO154790250
Citraconic anhydride 98%
Citraconic anhydride
Formula: C₅H₄O₃
Boiling Pt: 213 °C (760 mmHg)
Melting Pt: 6…7 °C
Density: 1,245 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4690 to 1.4730 (20°C, 589 nm)
Titration with NaOH 97.5 to 102.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR