3,5-Dimethoxybenzaldehyde 98%

Supplier: Thermo Fisher Scientific
Warning

155220100 155220500
ACRO155220100EA 113 EUR
ACRO155220100 ACRO155220500
3,5-Dimethoxybenzaldehyde 98%
3,5-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 151 °C (16 mmHg)
Melting Pt: 44…48 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003366
CAS Number: 7311-34-4
EINECS: 230-772-6

Order Now

Specification Test Results

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 44 to 48 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR