Dimethylammonium chloride 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dimethylamine hydrochloride

155270010 155270050 155275000 155270025
ACRO155270010EA 106 EUR
ACRO155270010 ACRO155270050 ACRO155275000 ACRO155270025
Dimethylammonium chloride 99%
Dimethylammonium chloride
Formula: C₂H₈ClN
MW: 81.54 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00012477
CAS Number: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤1 %
pH 5 to 6 (5 % in water)
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR