N,N,N',N'-Tetramethyl-1,3-propanediamine 99+%

Supplier: Thermo Fisher Scientific
Danger

155510100 155510500 155512500
ACRO155510100EA 36.9 EUR
ACRO155510100 ACRO155510500 ACRO155512500
N,N,N',N'-Tetramethyl-1,3-propanediamine 99+%
N,N,N',N'-Tetramethyl-1,3-propanediamine
Formula: C₇H₁₈N₂
Boiling Pt: 145…146 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0,78 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008337
CAS Number: 110-95-2
EINECS: 203-818-8
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4225 to 1.4245 (20°C, 589 nm)
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR