3-Nitrobenzoyl chloride 98%

Supplier: Thermo Fisher Scientific
Danger

155840050
ACRO155840050EA 25 EUR
ACRO155840050
3-Nitrobenzoyl chloride 98%
3-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 30…35 °C
Density: 1,42 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007247
CAS Number: 121-90-4
EINECS: 204-505-9
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance Brown Low melting solid
Infrared spectrum Conforms
Melting point 30 to 35 °C
Titration RCOCl ≥97.5 %
UV at 380 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0003 meq/g
Alkalinity ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR