2,3-Dimethyl-2-butene 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tetramethylethylene

156590250 156591000 156595000
ACRO156590250EA 71.8 EUR
ACRO156590250 ACRO156591000 ACRO156595000
2,3-Dimethyl-2-butene 98%
2,3-Dimethyl-2-butene
Formula: C₆H₁₂
Boiling Pt: 73 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0,7 g/cm³
Flash Pt: –16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR