Methylammonium chloride 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: MACl, Methylammonium chloride, Aminomethane hydrochloride, Methylamine hydrochloride

156780010 156782500 156785000 156780050 156780500
ACRO156780010EA 147 EUR
ACRO156780010 ACRO156782500 ACRO156785000 ACRO156780050 ACRO156780500
Methylammonium chloride 99%
Methylammonium chloride
Formula: CH₆ClN
MW: 67.52 g/mol
Boiling Pt: 225…230 °C (15 mmHg)
Melting Pt: 228…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00012849
CAS Number: 593-51-1
EINECS: 209-795-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 228 to 233 °C
Titration with HClO4 ≥98.5 %
Sulfate (SO4) ≤0.3 %
Iron (Fe) ≤0.005 %
Sodium (Na) ≤0.1 %
Heavy metals (as Pb) ≤0.005 %
Insoluble matter ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR