2-Methyl-3-buten-2-ol 97%

Supplier: Thermo Fisher Scientific
Danger

159670010 159672500
ACRO159670010EA 93.9 EUR
ACRO159670010 ACRO159672500
2-Methyl-3-buten-2-ol 97%
2-Methyl-3-buten-2-ol
Formula: C₅H₁₀O
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0,82 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00004470
CAS Number: 115-18-4
EINECS: 204-068-4
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20°C, 589 nm)
GC ≥96.0 %
Elemental Analysis 8.35 to 9.42 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR