2-Bromobenzoic acid 97%

Supplier: Thermo Fisher Scientific
Warning

160080250 160081000
ACRO160080250EA 39.7 EUR
ACRO160080250 ACRO160081000
2-Bromobenzoic acid 97%
2-Bromobenzoic acid
Formula: C₇H₅BrO₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 146…151 °C
Density: 1,929 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 145 to 150 °C
HPLC ≥96.0 %
Fines ≤8.5 % (< 0.5 cm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR