trans-Stilbene 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene

161041000 161045000
ACRO161041000EA 114 EUR
ACRO161041000 ACRO161045000
trans-Stilbene 96%
trans-Stilbene
Formula: C₁₄H₁₂
Boiling Pt: 305…307 °C (744 mmHg)
Melting Pt: 122…126 °C
Density: 0,9707 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064300
CAS Number: 103-30-0
EINECS: 203-098-5
UN: 3077
ADR: 9,III
Merck Index: 12,08972
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 122 to 126 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR