2-Chloro-3-nitrobenzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

161480050 161480250
ACRO161480050EA 178 EUR
ACRO161480050 ACRO161480250
2-Chloro-3-nitrobenzoic acid 98%
2-Chloro-3-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Melting Pt: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2
EINECS: 223-590-3

Order Now

Specification Test Results

Appearance White to yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 183 to 188 °C
HPLC ≥97.5 %
GC ≥92 %
Specific optical rotation -31° to -35° (20 °C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR