Benzyltriethylammonium chloride 98%

Supplier: Thermo Fisher Scientific
Warning

164021000 164025000 164020025
ACRO164021000EA 32.8 EUR
ACRO164021000 ACRO164025000 ACRO164020025
Benzyltriethylammonium chloride 98%
Benzyltriethylammonium chloride
Formula: C₁₃H₂₂ClN
Melting Pt: 185 °C (dec.)
Flash Pt: >275 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Water ≤2.0 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR