trans-Benzylideneacetone 99%

Supplier: Thermo Fisher Scientific
Warning

164525000 164520500 164521000
ACRO164525000EA 328 EUR
ACRO164525000 ACRO164520500 ACRO164521000
trans-Benzylideneacetone 99%
trans-Benzylideneacetone
Formula: C₁₀H₁₀O
Boiling Pt: 260…262 °C (760 mmHg)
Melting Pt: 38,5…41 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00008779
CAS Number: 1896-62-4
EINECS: 217-587-6

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals or solid
Infrared spectrum Conforms
Melting point 38.5 to 41 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR