Betaine hydrochloride 99%, Extra pure meets analytical specification of USP

Supplier: Thermo Fisher Scientific
Warning

164550010 164550025 164552500
ACRO164550010EA 164 EUR
ACRO164550010 ACRO164550025 ACRO164552500
Betaine hydrochloride 99%, Extra pure meets analytical specification of USP
Betaine hydrochloride
Formula: C₅H₁₂ClNO₂
Melting Pt: 241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011903
CAS Number: 590-46-5
EINECS: 209-683-1

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms (identification A)
Water ≤0.5 %
Identification B Passes test
Residue after ignition ≤0.1 %
pH 0.8 to 1.2 (1 in 4 solution)
Titration with HClO4 98.5 to 100.5 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR