Acrylamide 99+% for biochemistry, for electrophoresis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Prop-2-enamide

164851000 164855000 164850025
ACRO164851000EA 40.7 EUR
ACRO164851000 ACRO164855000 ACRO164850025
Acrylamide 99+% for biochemistry, for electrophoresis
Acrylamide (monomer)
Formula: C₃H₅NO
Boiling Pt: 125 °C (25 mmHg)
Melting Pt: 82…86 °C
Density: 1,122 g/cm³
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.2 % (K.F.)
GC ≥99.0 %
pH 5.5 to 6.5 (10 % in 0.1M NaCl at 25°C)
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Acrylic acid ≤0.002 %
Insoluble matter ≤0.005 % (in water)
Conductivity ≤5 µmho (40% aq. soln.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR