Dimethyl sulphoxide-[D₆] 99.9 atom % D for NMR spectroscopy

Supplier: Thermo Fisher Scientific

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

166290100 166290500 166291000
ACRO166290100EA 111 EUR
ACRO166290100 ACRO166290500 ACRO166291000
Dimethyl sulphoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189
Melting Pt: 20.2
Density: 1.19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (K.F.)
D-Enrichment ≥99.85 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR