4-Bromo-α,α,α-trifluorotoluene 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

166670050 166670250
ACRO166670050EA 60 EUR
ACRO166670050 ACRO166670250
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 154…155 °C (760 mmHg)
Density: 1.6 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4745 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR