1,4-Naphthoquinone, (max. 6% H₂O) 99%

Supplier: Thermo Fisher Scientific
Danger

166711000 166715000 166710025
ACRO166711000EA 36.2 EUR
ACRO166711000 ACRO166715000 ACRO166710025
1,4-Naphthoquinone, (max. 6% H₂O) 99%
1,4-Naphthoquinone
Formula: C₁₀H₆O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 128,5 °C
Density: 1,422 g/cm³ (20 °C)
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2928
ADR: 6.1,I
Merck Index: 12,06482

Order Now

Specification Test Results

Appearance Khaki Powder
Infrared spectrum Conforms
Water ≤6 % (K.F.)
GC ≥99 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR