Capric acid 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Decylic acid, Decanoic acid

167271000 167275000 167270010 167270025
ACRO167271000EA 53.3 EUR
ACRO167271000 ACRO167275000 ACRO167270010 ACRO167270025
Capric acid 99%
Capric acid
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268…270 °C (760 mmHg)
Melting Pt: 30…32 °C
Density: 0.89 g/cm³
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Order Now

Specification Test Results

Appearance Colorless or white to cream Crystals and/or chunks or fused solid or liquid
Infrared spectrum Conforms
Melting point 29 to 34 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR