5-Bromo-o-anisaldehyde 99%

Supplier: Thermo Fisher Scientific
Warning

167520250 167520050 167521000
ACRO167520250EA 50.8 EUR
ACRO167520250 ACRO167520050 ACRO167521000
5-Bromo-o-anisaldehyde 99%
5-Bromo-o-anisaldehyde
Formula: C₈H₇BrO₂
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3

Order Now

Specification Test Results

Appearance Off-white to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 115 to 119 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR