Cyclohexane 99+% for spectroscopy

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

167740010 167740025 167745000
ACRO167740010EA 121 EUR
ACRO167740010 ACRO167740025 ACRO167745000
Cyclohexane 99+% for spectroscopy
Cyclohexane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 6.5 °C
Density: 0.77 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 %
GC ≥99.0 %
UV at 260 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR