Ethyl acetate 99.5+% for spectroscopy

Supplier: Thermo Fisher Scientific
Danger

167810010 167810025 167815000
ACRO167810010EA 65.3 EUR
ACRO167810010 ACRO167810025 ACRO167815000
Ethyl acetate 99.5+% for spectroscopy
Ethyl acetate
Formula: CH₃COOC₂H₅
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83,5 °C
Density: 0,902 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.50 %
UV at 325 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR