2-(2-Pyridyl)benzimidazole 99%

Supplier: Thermo Fisher Scientific
Warning

167950050 167950100
ACRO167950050EA 78.4 EUR
ACRO167950050 ACRO167950100
2-(2-Pyridyl)benzimidazole 99%
2-(2-Pyridyl)benzimidazole

Appearance: Yellow to brown Powder

Formula: C₁₂H₉N₃
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005586
CAS Number: 1137-68-4

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR