2-Fluoro-5-nitroaniline 98%

Supplier: Thermo Fisher Scientific
Warning

168390250 168391000 168390050
ACRO168390250EA 71.4 EUR
ACRO168390250 ACRO168391000 ACRO168390050
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
Melting Pt: 99…103 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Ochre to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR