Cyclohexanecarbonyl chloride 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride

169050250 169051000
ACRO169050250EA 30 EUR
ACRO169050250 ACRO169051000
Cyclohexanecarbonyl chloride 98%
Cyclohexanecarbonyl chloride
Formula: C₇H₁₁ClO
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 29 °C
Density: 1,09 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00001456
CAS Number: 2719-27-9
EINECS: 220-322-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4700 (20°C, 589 nm)
GC ≥97.5 %
Titration RCOCl ≥97.5 %
Titration RCOOH ≤2 %
Free acid (HCl) Traces


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR