ω-Bromo-4-methylacetophenone 97%

Supplier: Thermo Fisher Scientific
Warning

170390050 170390250 170391000
ACRO170390050EA 49.2 EUR
ACRO170390050 ACRO170390250 ACRO170391000
ω-Bromo-4-methylacetophenone 97%
ω-Bromo-4-methylacetophenone
Formula: C₉H₉BrO
Boiling Pt: 105 °C (0,1 -rr)
Melting Pt: 52…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000203
CAS Number: 619-41-0
EINECS: 210-595-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR