Sodium iodoacetate 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium 2-iodoacetate, Iodoacetic Acid, Sodium Salt

170970250 170971000
ACRO170970250EA 128 EUR
ACRO170970250 ACRO170971000
Sodium iodoacetate 99%
Sodium iodoacetate
Formula: C₂H₂INaO₂
Melting Pt: 208…210 °C
Flash Pt: 208 °C
Storage Temperature: Freezer
MDL Number: MFCD00002686
CAS Number: 305-53-3
EINECS: 206-165-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥96.0 %
Stabilizer hydroquinone: approx. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR