meso-Tetraphenylporphine 97%

Supplier: Thermo Fisher Scientific
Warning

171210010 171210050 171210250
ACRO171210010EA 64.9 EUR
ACRO171210010 ACRO171210050 ACRO171210250
meso-Tetraphenylporphine 97%
meso-Tetraphenylporphine
Formula: C₄₄H₃₀N₄
Melting Pt: >300 °C
Density: 1,276 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9

Order Now

Specification Test Results

Appearance Blue to purple Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Impurity ≤3 % Chlorin


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR