5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%

Supplier: Thermo Fisher Scientific

173090050 173090250
ACRO173090050EA 91.3 EUR
ACRO173090050 ACRO173090250
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride

CAS No.: 72-40-2

Formula: C₄H₇ClN₄O
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012704
CAS Number: 72-40-2

Order Now

Specification Test Results

Appearance White to light beige to light grey Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Solubility (5 % in water) Clear
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Peroxides ≤0.0075 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR