Tetrabutylammonium chloride 95% tech. contains also bromide

Supplier: Thermo Fisher Scientific
Warning

174120050 174120250 174121000 174125000
ACRO174120050EA 35.3 EUR
ACRO174120050 ACRO174120250 ACRO174121000 ACRO174125000
Tetrabutylammonium chloride 95% tech. contains also bromide
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
Melting Pt: 83…86 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00011635
CAS Number: 1112-67-0

Order Now

Specification Test Results

Appearance White to brown Crystalline solid
Infrared spectrum Conforms
Titration Argentometric ≥94 % (on dry substance)
Water ≤4 %
Bromide (Br) ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR