Triethyl-2-phosphonopropionate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate

174360100 174360500
ACRO174360100EA 65.7 EUR
ACRO174360100 ACRO174360500
Triethyl-2-phosphonopropionate 98%
Triethyl 2-phosphonopropionate
Formula: C₉H₁₉O₅P
MW: 238.22 g/mol
Boiling Pt: 143…144 °C (12 mmHg)
Density: 1.11 g/cm³
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009159
CAS Number: 3699-66-9
EINECS: 223-033-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR