(3-Mercaptopropyl)trimethoxysilane 85% tech.

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol

174650500 174652500
ACRO174650500EA 63.4 EUR
ACRO174650500 ACRO174652500
(3-Mercaptopropyl)trimethoxysilane 85% tech.
(3-Mercaptopropyl)trimethoxysilane
Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1,05 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00004901
CAS Number: 4420-74-0
EINECS: 224-588-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20°C, 589 nm)
GC ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR