tert-Butyl methyl ether 99%, pure

Supplier: Thermo Fisher Scientific
Danger

177040010 177040025 177042500 177040250 177040100
ACRO177040010EA 46 EUR
ACRO177040010 ACRO177040025 ACRO177042500 ACRO177040250 ACRO177040100
tert-Butyl methyl ether 99%, pure
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
Boiling Pt: 54…56 °C (760 mmHg)
Melting Pt: –110 °C
Density: 0,7404 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3675 to 1.3695 (20°C, 589 nm)
Water ≤0.01 % (Coulometric)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR