D-(+)-Sucrose 99.7% for biochemistry

Supplier: Thermo Fisher Scientific

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

177140010 177140050 177140250 177142500
ACRO177140010EA 73.4 EUR
ACRO177140010 ACRO177140050 ACRO177140250 ACRO177142500
D-(+)-Sucrose 99.7% for biochemistry
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 190…192 °C (dec.)
Density: 1,29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Coarse crystals
Infrared spectrum Conforms
Loss on drying ≤0.1 %
Sulfated ash ≤0.02 %
HPLC ≥99.7 % (ELSD)
pH 6.5 to 7.5 (10 % solution)
Specific optical rotation (20°C, 589 nm) (c=26, H2O) > +65.9°
Turbidity ≤3 NTU (in water)
Sulfite (as SO2) ≤15 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.0007 % (in water)
Reducing sugars ≤0.04 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR