Sodium alginate

Supplier: Thermo Fisher Scientific

Synonyms: Alginic acid sodium salt, Algin

177770050 177772500 177775000 177770025
ACRO177770050EA 31.9 EUR
ACRO177770050 ACRO177772500 ACRO177775000 ACRO177770025
Sodium alginate
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Specification Test Results

Appearance Off-white to yellow-brown Powder
Infrared spectrum Conforms
Loss on drying ≤15.0 % (100 mg, 105°C, 4 hrs)
Sulfated ash 30.0 % to 36.0 % (on dried substance)
Viscosity 350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
pH 5.5 to 8.0 (1 % at 20°C)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR