Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Danger

180130010 180130050 180130100
ACRO180130010EA 89.8 EUR
ACRO180130010 ACRO180130050 ACRO180130100
Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Tetrahydrofuran-[D8]
Formula: C₄D₈O
Boiling Pt: 65…66 °C (760 mmHg)
Melting Pt: –106 °C
Density: 0,98 g/cm³
Flash Pt: –21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.06 % (Coulometric)
D-Enrichment ≥99.4 %
Titration with HClO4 ≥98.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR