2,6-Dimethoxybenzonitrile 97%

Supplier: Thermo Fisher Scientific
Warning

181720100 181720500
ACRO181720100EA 43.4 EUR
ACRO181720100 ACRO181720500
2,6-Dimethoxybenzonitrile 97%
2,6-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00001788
CAS Number: 16932-49-3
EINECS: 241-000-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 119 to 123 °C
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR