3,4-Diaminobenzophenone 99%

Supplier: Thermo Fisher Scientific
Warning

184800250
ACRO184800250EA 52.2 EUR
ACRO184800250
3,4-Diaminobenzophenone 99%
3,4-Diaminobenzophenone
Formula: C₁₃H₁₂N₂O
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: 116…119 °C
MDL Number: MFCD00007727
CAS Number: 39070-63-8
EINECS: 254-273-8

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder
Infrared spectrum Conforms
Melting point 115 to 119 °C
Titration with HClO4 ≥98.5 %
HPLC ≥95.0 %
Specific optical rotation (Dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR