Boc-ON 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(tert-BOC-Oxyimino)-2-phenylacetonitrile, N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide, 2-(Boc-oxyimino)-2-phenylacetonitrile, 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile, [(cyano-phenyl-methylidene)amino] tert-butyl carbonate

185890250 185890050 185891000
ACRO185890250EA 117 EUR
ACRO185890250 ACRO185890050 ACRO185891000
Boc-ON 99%
Boc-ON

Appearance: White to light beige Crystalline powder

Formula: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
Melting Pt: 86…89 °C
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001863
CAS Number: 58632-95-4
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 89 °C
Water ≤0.1 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation +116° to +125° (20 °C, 589 nm) (1 % in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR