Hexafluorophosphoric acid 60% (w/w) in water

Supplier: Thermo Fisher Scientific
Danger

191110250 191115000
ACRO191110250EA 38.8 EUR
ACRO191110250 ACRO191115000
Hexafluorophosphoric acid 60% (w/w) in water
Hexafluorophosphoric acid
Formula: HF₆P
Density: 1,75 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011350
CAS Number: 16940-81-1
EINECS: 241-006-5
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to yellow-green Solution
Assay Titrimetry ≥55.0 %
Additional info the presence of these hydrolysis products may rise the strength to over 70%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR