Germanium 99.999% (trace metals basis), powder

Supplier: Thermo Fisher Scientific
Warning

193110100 193110500
ACRO193110100EA 242 EUR
ACRO193110100 ACRO193110500
Germanium 99.999% (trace metals basis), powder
Germanium
Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937 °C
Density: 5,35 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III
Merck Index: 12,04415

Order Now

Specification Test Results

Appearance Grey to black Glistening crystalline powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR