Mercury (II) oxide, red 99+% ACS

Supplier: Thermo Fisher Scientific
Danger

193510250 193511000
ACRO193510250EA 48.9 EUR
ACRO193510250 ACRO193511000
Mercury (II) oxide, red 99+% ACS
Mercury (II) oxide
Formula: HgO
Boiling Pt: 356,6 °C (1013 hPa)
Melting Pt: 500 °C (dec.)
Density: 11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
UN: 1641
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Orange-red to red Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤0.025 %
Sulfate (SO4) ≤0.015 %
Iron (Fe) ≤0.005 %
Insoluble matter ≤0.03 % (in dilute HCl)
Residue after reduction ≤0.025 %
Nitrogen compounds (as N) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR