Potassium cyanide 97+% for analysis

Supplier: Thermo Fisher Scientific
Danger

196605000 196600025 196601000 196602500
ACRO196605000EA 159 EUR
ACRO196605000 ACRO196600025 ACRO196601000 ACRO196602500
Potassium cyanide 97+% for analysis
Potassium cyanide
Formula: KCN
Boiling Pt: 1625 °C
Melting Pt: 634 °C
Density: 1,55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 13,07709

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥97.0 %
Chloride (Cl) ≤500 ppm
Thiocyanate (SCN) ≤100 ppm
Phosphate (PO4) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤100 ppm
Lead (Pb) ≤5 ppm
Sodium (Na) ≤5000 ppm
Sulfur (S) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR