Sodium nitrite 99% contains an anticaking reagent, extra pure

Supplier: Thermo Fisher Scientific
Danger

196620025 196620010 196620100
ACRO196620025EA 90.2 EUR
ACRO196620025 ACRO196620010 ACRO196620100
Sodium nitrite 99% contains an anticaking reagent, extra pure
Sodium nitrite
Formula: NaNO₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 271 °C
Density: 2,168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
UN: 1500
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Water ≤0.3 %
Titration ≥98.5 %
Anticaking agent ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR