Sodium hexafluorophosphate 98.5+%, pure

Supplier: Thermo Fisher Scientific
Danger

196820500 196820100 196822500
ACRO196820500EA 310 EUR
ACRO196820500 ACRO196820100 ACRO196822500
Sodium hexafluorophosphate 98.5+%, pure
Sodium hexafluorophosphate
Formula: F₆NaP
Melting Pt: 200 °C
Density: 2,505 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light grey Crystalline powder
Assay ≥98.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR